siRNA Details
Gene Symbol: CYP4Z2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002788.2 | 1, 2 | 226 | |||
GenBank mRNA | AK097373.1 | - | 1, 2 | 226 | ||
AY262057.1 | - | 1, 2 | 188 | |||
AY696295.1 | - | 1, 2 | 185 | |||
DA998990.1 | - | 1, 2 | 227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
