siRNA Details
Gene Symbol: USP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015306.2 | NP_056121.2 | 44 | 5443 | ||
XM_005270690.3 | XP_005270747.1 | 31 | 3484 | |||
XM_017000831.1 | XP_016856320.1 | 45 | 5508 | |||
XM_017000832.1 | XP_016856321.1 | 45 | 5505 | |||
XM_017000833.1 | XP_016856322.1 | 45 | 5000 | |||
XM_017000834.1 | XP_016856323.1 | 44 | 4940 | |||
GenBank mRNA | AB028980.1 | - | 3 | 262 | ||
AK127075.1 | - | 27 | 3188 | |||
AL832370.1 | - | 6 | 669 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
