siRNA Details
Gene Symbol: USP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015306.2 | NP_056121.2 | 63 | 7662 | ||
XM_005270690.3 | XP_005270747.1 | 50 | 5703 | |||
XM_017000831.1 | XP_016856320.1 | 64 | 7727 | |||
XM_017000832.1 | XP_016856321.1 | 64 | 7724 | |||
XM_017000833.1 | XP_016856322.1 | 64 | 7219 | |||
XM_017000834.1 | XP_016856323.1 | 63 | 7159 | |||
GenBank mRNA | BC029660.1 | - | 8 | 715 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
