siRNA Details
Gene Symbol: ZSWIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020883.1 | NP_065934.1 | 2 | 1011 | ||
XM_011541861.2 | XP_011540163.1 | 2 | 1019 | |||
XM_017001913.1 | XP_016857402.1 | 1 | 97 | |||
GenBank mRNA | AB040944.1 | - | 2 | 989 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
