siRNA Details
Gene Symbol: KIAA1522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198972.1 | NP_001185901.1 | 7 | 3581 | ||
NM_001198973.1 | NP_001185902.1 | 5 | 905 | |||
NM_020888.2 | NP_065939.2 | 7 | 3666 | |||
XM_006710791.3 | XP_006710854.1 | 7 | 3600 | |||
XM_017001916.1 | XP_016857405.1 | 5 | 3264 | |||
GenBank mRNA | AB040955.1 | - | 7 | 3548 | ||
AL713671.1 | - | 2 | 1321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
