siRNA Details
Gene Symbol: MYSM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085487.2 | NP_001078956.1 | 7, 8 | 522 | ||
XM_006710314.3 | XP_006710377.1 | 7, 8 | 538 | |||
XM_011540573.2 | XP_011538875.1 | 8, 9 | 502 | |||
XM_011540574.2 | XP_011538876.1 | 7, 8 | 770 | |||
XM_017000195.1 | XP_016855684.1 | 8, 9 | 574 | |||
XM_017000196.1 | XP_016855685.1 | 5, 6 | 424 | |||
XM_017000197.1 | XP_016855686.1 | 3, 4 | 202 | |||
GenBank mRNA | AB067502.1 | - | 3, 4 | 548 | ||
AK126835.1 | - | 1, 2 | 71 | |||
BX648168.1 | - | 4, 5 | 258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
