siRNA Details
Gene Symbol: ADGRL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022159.3 | NP_071442.2 | 9 | 1254 | ||
GenBank mRNA | AB032017.1 | - | 9 | 1215 | ||
AF192403.1 | - | 7 | 915 | |||
AK172750.1 | - | 6 | 733 | |||
AK313738.1 | - | 9 | 1253 | |||
AY358360.1 | - | 9 | 1121 | |||
BC025721.1 | - | 9 | 1120 | 5229055 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
