siRNA Details
Gene Symbol: LOC105378803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003080.1 | XP_016858569.1 | 2 | 298 | ||
XM_017003081.1 | XP_016858570.1 | 2 | 3543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
