siRNA Details
Gene Symbol: VANGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020335.2 | NP_065068.1 | 8 | 3459 | ||
XM_005245357.1 | XP_005245414.1 | 8 | 3266 | |||
XM_011509804.1 | XP_011508106.1 | 8 | 3314 | |||
GenBank mRNA | AB033041.1 | - | 7 | 3135 | ||
AL137558.1 | - | 1 | 611 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
