siRNA Details
Gene Symbol: KIF26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018012.3 | NP_060482.2 | 10 | 2598 | ||
GenBank mRNA | AK128806.1 | - | 1 | 91 | ||
AK293964.1 | - | 10 | 2513 | |||
AK297303.1 | - | 10 | 2594 | |||
AY923834.1 | - | 10 | 2599 | |||
BC035896.1 | - | 7 | 1119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
