siRNA Details
Gene Symbol: KIF26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018012.3 | NP_060482.2 | 8 | 2181 | ||
GenBank mRNA | AK293964.1 | - | 8 | 2096 | ||
AK297303.1 | - | 8 | 2177 | |||
AY923834.1 | - | 8 | 2182 | |||
BC035896.1 | - | 5 | 702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
