siRNA Details
Gene Symbol: KIF26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018012.3 | NP_060482.2 | 15 | 7069 | ||
GenBank mRNA | AK001019.1 | - | 4 | 2008 | ||
AK125187.1 | - | 8 | 2524 | |||
AY923834.1 | - | 15 | 7070 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
