siRNA Details
Gene Symbol: PKDCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138370.2 | NP_612379.2 | 7 | 2004 | ||
GenBank mRNA | BC007901.2 | - | 5 | 959 | ||
BC062988.1 | - | 5 | 959 | |||
BC094697.1 | - | 7 | 1412 | 30529646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
