siRNA Details
Gene Symbol: FBXO41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080410.2 | NP_001073879.2 | 12 | 3383 | ||
XM_005264158.3 | XP_005264215.1 | 10 | 2302 | |||
XM_006711950.2 | XP_006712013.1 | 17 | 4367 | |||
XM_011532562.1 | XP_011530864.1 | 13 | 3815 | |||
XM_011532563.2 | XP_011530865.1 | 11 | 2740 | |||
XM_017003421.1 | XP_016858910.1 | 14 | 6464 | |||
XM_017003422.1 | XP_016858911.1 | 17 | 6109 | |||
XM_017003423.1 | XP_016858912.1 | 17 | 5490 | |||
GenBank mRNA | AB075820.1 | - | 12 | 3221 | ||
AK055121.1 | - | 1 | 389 | |||
AK095028.1 | - | 3 | 1054 | |||
AK127503.1 | - | 9 | 1864 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
