siRNA Details
Gene Symbol: CFAP221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006712353.3 | XP_006712416.1 | 19 | 2080 | ||
XM_017003550.1 | XP_016859039.1 | 19 | 2080 | |||
XM_017003551.1 | XP_016859040.1 | 19 | 2207 | |||
XM_017003552.1 | XP_016859041.1 | 19 | 2093 | |||
XM_017003553.1 | XP_016859042.1 | 19 | 2426 | |||
XM_017003554.1 | XP_016859043.1 | 18 | 1922 | |||
XM_017003555.1 | XP_016859044.1 | 18 | 1922 | |||
XM_017003556.1 | XP_016859045.1 | 18 | 1925 | |||
XM_017003557.1 | XP_016859046.1 | 17 | 1855 | |||
XM_017003558.1 | XP_016859047.1 | 19 | 2080 | |||
XM_017003559.1 | XP_016859048.1 | 16 | 3051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
