siRNA Details
Gene Symbol: XIRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079810.3 | NP_001073278.1 | 10 | 1550 | ||
NM_001199143.1 | NP_001186072.1 | 11 | 1649 | |||
NM_001199144.1 | NP_001186073.1 | 9 | 10527 | |||
NM_001199145.1 | NP_001186074.1 | 8 | 1148 | |||
NM_152381.5 | NP_689594.4 | 11 | 10929 | |||
XM_017003309.1 | XP_016858798.1 | 9 | 10424 | |||
GenBank mRNA | AK056582.1 | - | 3 | 2157 | ||
AK097519.1 | - | 10 | 1650 | |||
AK122889.1 | - | 9 | 989 | |||
AL831970.2 | - | 3 | 3131 | |||
AL831985.1 | - | 3 | 2791 | |||
AL832011.1 | - | 3 | 3415 | |||
AL832336.1 | - | 3 | 3456 | |||
AL832360.1 | - | 3 | 3036 | |||
AL832375.1 | - | 3 | 2466 | |||
AL832382.1 | - | 3 | 2983 | |||
AL832452.2 | - | 4 | 3504 | |||
AL833291.2 | - | 4 | 4747 | |||
BC022888.1 | - | 3 | 792 | |||
BX647448.1 | - | 3 | 4479 | |||
BX647645.1 | - | 3 | 3150 | |||
BX647844.1 | - | 3 | 4330 | |||
BX647845.1 | - | 3 | 3088 | |||
BX647848.1 | - | 3 | 4968 | |||
EF119711.1 | - | 9 | 10527 | |||
EF119717.1 | - | 8 | 1148 | |||
EF119718.1 | - | 12 | 1749 | |||
EF119719.1 | - | 10 | 1651 | |||
EF119720.1 | - | 10 | 1550 | |||
EF119721.1 | - | 11 | 1750 | |||
EF119722.1 | - | 11 | 1649 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
