siRNA Details
Gene Symbol: UBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172070.3 | NP_742067.3 | 8 | 1271 | ||
XM_005246299.4 | XP_005246356.1 | 8 | 1316 | |||
XM_005246300.4 | XP_005246357.1 | 8 | 1271 | |||
XM_006712268.3 | XP_006712331.1 | 8 | 1313 | |||
XM_006712269.3 | XP_006712332.1 | 8 | 1271 | |||
XM_006712270.3 | XP_006712333.2 | 7 | 1172 | |||
XM_006712271.3 | XP_006712334.1 | 8 | 1312 | |||
XM_011510604.2 | XP_011508906.1 | 8 | 1271 | |||
XM_017003356.1 | XP_016858845.1 | 8 | 1271 | |||
XM_017003357.1 | XP_016858846.1 | 8 | 1271 | |||
XM_017003358.1 | XP_016858847.1 | 8 | 1271 | |||
XM_017003359.1 | XP_016858848.1 | 8 | 1271 | |||
GenBank mRNA | DR731153.1 | - | 6 | 453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
