siRNA Details
Gene Symbol: SLC9A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011552.3 | NP_001011552.2 | 8 | 2047 | ||
XM_011511158.1 | XP_011509460.1 | 8 | 1981 | |||
GenBank mRNA | CR627411.1 | - | 8 | 2048 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
