siRNA Details
Gene Symbol: C1orf189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010979.2 | NP_001010979.1 | 3 | 191 | ||
GenBank mRNA | BC127710.1 | - | 3 | 190 | 40127224 | |
BC130509.1 | - | 3 | 249 | 40146540 | ||
HY025946.1 | - | 3 | 191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
