siRNA Details
Gene Symbol: LINC01116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040001.2 | 3 | 387 | |||
GenBank mRNA | BC013438.1 | - | 3 | 744 | ||
BC013588.1 | - | 1 | 56 | |||
BE908176.1 | - | 3 | 361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
