siRNA Details
Gene Symbol: EIF4E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134649.2 | NP_001128121.1 | 7 | 702 | ||
NM_001134650.1 | NP_001128122.1 | 7 | 578 | |||
NM_001134651.1 | NP_001128123.1 | 6 | 569 | |||
NM_001282886.1 | NP_001269815.1 | 7 | 695 | |||
NM_173359.4 | NP_775495.1 | 7 | 663 | |||
XM_011533651.2 | XP_011531953.1 | 4 | 422 | |||
XM_017006278.1 | XP_016861767.1 | 8 | 709 | |||
XM_017006279.1 | XP_016861768.1 | 8 | 696 | |||
XM_017006280.1 | XP_016861769.1 | 8 | 843 | |||
XM_017006281.1 | XP_016861770.1 | 6 | 571 | |||
XM_017006282.1 | XP_016861771.1 | 8 | 651 | |||
GenBank mRNA | AK126999.1 | - | 6 | 451 | ||
AK313656.1 | - | 7 | 663 | |||
BC031289.2 | - | 5 | 386 | 5299499 | ||
BC068443.1 | - | 7 | 695 | 4820931 | ||
BX648727.1 | - | 3 | 183 | |||
DA227887.1 | - | 7 | 553 | |||
DR159502.1 | - | 6 | 568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
