siRNA Details
Gene Symbol: SRGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170637.3 | NP_001164108.1 | 4, 5 | 1033 | ||
NM_001300952.1 | NP_001287881.1 | 4, 5 | 1033 | |||
NM_015326.4 | NP_056141.2 | 4, 5 | 1033 | |||
XM_005277510.2 | XP_005277567.1 | 5, 6 | 1220 | |||
XM_005277511.2 | XP_005277568.1 | 5, 6 | 1220 | |||
XM_005277514.1 | XP_005277571.1 | 5, 6 | 1220 | |||
XM_005277515.1 | XP_005277572.1 | 5, 6 | 1220 | |||
XM_011509354.1 | XP_011507656.1 | 5, 6 | 1220 | |||
XM_011509355.1 | XP_011507657.1 | 5, 6 | 1220 | |||
XM_011509356.2 | XP_011507658.1 | 5, 6 | 1220 | |||
XM_011509359.1 | XP_011507661.1 | 5, 6 | 1220 | |||
XM_017000838.1 | XP_016856327.1 | 3, 4 | 864 | |||
XM_017000839.1 | XP_016856328.1 | 3, 4 | 894 | |||
XM_017000840.1 | XP_016856329.1 | 5, 6 | 509 | |||
XM_017000841.1 | XP_016856330.1 | 5, 6 | 1220 | |||
GenBank mRNA | AB007925.1 | - | 4, 5 | 563 | ||
AK293335.1 | - | 1, 2 | 69 | |||
AK295845.1 | - | 4, 5 | 789 | |||
BC132872.1 | - | 4, 5 | 561 | 40146894 | ||
BC132874.1 | - | 4, 5 | 561 | 40146896 | ||
BC144343.1 | - | 4, 5 | 561 | 9052863 | ||
S68466.1 | - | 1, 2 | 49 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
