siRNA Details
Gene Symbol: SRGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170637.3 | NP_001164108.1 | 22 | 4972 | ||
NM_015326.4 | NP_056141.2 | 22 | 4975 | |||
XM_005277510.2 | XP_005277567.1 | 23 | 5162 | |||
XM_005277511.2 | XP_005277568.1 | 23 | 5159 | |||
XM_011509354.1 | XP_011507656.1 | 24 | 5201 | |||
XM_011509355.1 | XP_011507657.1 | 24 | 5198 | |||
XM_011509356.2 | XP_011507658.1 | 25 | 5296 | |||
XM_017000838.1 | XP_016856327.1 | 22 | 4845 | |||
XM_017000839.1 | XP_016856328.1 | 22 | 4875 | |||
XM_017000840.1 | XP_016856329.1 | 24 | 4490 | |||
GenBank mRNA | AB007925.1 | - | 22 | 4507 | ||
BC132872.1 | - | 22 | 4497 | 40146894 | ||
BC132874.1 | - | 22 | 4497 | 40146896 | ||
BC144343.1 | - | 22 | 4494 | 9052863 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
