siRNA Details
Gene Symbol: DNAH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291661.1 | NP_001278590.1 | 26 | 4108 | ||
XM_011533466.1 | XP_011531768.1 | 26 | 4105 | |||
XM_011533467.1 | XP_011531769.1 | 26 | 4072 | |||
XM_011533468.1 | XP_011531770.1 | 26 | 4105 | |||
XM_011533469.1 | XP_011531771.1 | 26 | 4105 | |||
XM_011533471.2 | XP_011531773.1 | 26 | 4105 | |||
XM_017005860.1 | XP_016861349.1 | 26 | 4105 | |||
XM_017005861.1 | XP_016861350.1 | 25 | 4788 | |||
XM_017005862.1 | XP_016861351.1 | 5 | 680 | |||
GenBank mRNA | U53532.1 | - | 4 | 517 | ||
U61739.1 | - | 1 | 101 | |||
Z83802.1 | - | 1 | 102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
