siRNA Details
Gene Symbol: DNAH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291661.1 | NP_001278590.1 | 26 | 4030 | ||
XM_011533466.1 | XP_011531768.1 | 26 | 4027 | |||
XM_011533467.1 | XP_011531769.1 | 26 | 3994 | |||
XM_011533468.1 | XP_011531770.1 | 26 | 4027 | |||
XM_011533469.1 | XP_011531771.1 | 26 | 4027 | |||
XM_011533471.2 | XP_011531773.1 | 26 | 4027 | |||
XM_017005860.1 | XP_016861349.1 | 26 | 4027 | |||
XM_017005861.1 | XP_016861350.1 | 25 | 4710 | |||
XM_017005862.1 | XP_016861351.1 | 5 | 602 | |||
GenBank mRNA | U53532.1 | - | 4 | 439 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
