siRNA Details
Gene Symbol: DCLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033403.1 | NP_208382.1 | 3 | 1969 | ||
XM_011534167.2 | XP_011532469.1 | 3 | 1928 | |||
XM_011534168.2 | XP_011532470.1 | 3 | 1969 | |||
GenBank mRNA | AB051552.1 | - | 2 | 1359 | ||
AK122988.1 | - | 2 | 1288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
