siRNA Details
Gene Symbol: TCF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175769.2 | NP_786951.1 | 2, 3 | 515 | ||
XM_005264159.4 | XP_005264216.1 | 2, 3 | 697 | |||
GenBank mRNA | BC137191.1 | - | 2, 3 | 515 | 9021188 | |
BC137192.1 | - | 2, 3 | 515 | 9021189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
