siRNA Details
Gene Symbol: TCF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175769.2 | NP_786951.1 | 3 | 616 | ||
GenBank mRNA | BC137191.1 | - | 3 | 616 | 9021188 | |
BC137192.1 | - | 3 | 616 | 9021189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
