siRNA Details
Gene Symbol: C2orf16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032266.3 | NP_115642.3 | 1 | 3597 | ||
GenBank mRNA | AL136898.1 | - | 1 | 457 | ||
AL834384.1 | - | 1 | 131 | |||
BC115376.1 | - | 1 | 456 | |||
BC115377.1 | - | 1 | 456 | |||
BC140828.1 | - | 1 | 3593 | 9021696 | ||
CR749335.1 | - | 1 | 3597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
