siRNA Details
Gene Symbol: PLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170585.1 | NP_001164056.1 | 24 | 1664 | ||
NM_153021.4 | NP_694566.4 | 25 | 1697 | |||
XM_011532579.2 | XP_011530881.1 | 27 | 2004 | |||
XM_011532581.2 | XP_011530883.1 | 27 | 1967 | |||
XM_011532584.2 | XP_011530886.1 | 27 | 2004 | |||
XM_011532588.2 | XP_011530890.1 | 27 | 2006 | |||
XM_011532589.2 | XP_011530891.1 | 27 | 2001 | |||
XM_011532590.2 | XP_011530892.1 | 27 | 2006 | |||
XM_011532591.2 | XP_011530893.1 | 27 | 2003 | |||
XM_011532593.2 | XP_011530895.1 | 23 | 1579 | |||
XM_011532601.2 | XP_011530903.1 | 13 | 992 | |||
XM_011532602.2 | XP_011530904.1 | 12 | 964 | |||
XM_011532603.2 | XP_011530905.1 | 12 | 1041 | |||
XM_011532606.2 | XP_011530908.1 | 27 | 2026 | |||
XM_011532607.2 | XP_011530909.1 | 27 | 2026 | |||
XM_011532608.2 | XP_011530910.1 | 27 | 2024 | |||
XM_017003432.1 | XP_016858921.1 | 26 | 1973 | |||
XM_017003433.1 | XP_016858922.1 | 26 | 1939 | |||
XM_017003434.1 | XP_016858923.1 | 27 | 2008 | |||
GenBank mRNA | AK055428.1 | - | 6 | 380 | ||
BC065041.1 | - | 12 | 783 | |||
BC150633.1 | - | 25 | 1697 | 9057003 | ||
BC153864.1 | - | 24 | 1616 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
