siRNA Details
Gene Symbol: WDR43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015131.2 | NP_055946.1 | 5 | 717 | ||
GenBank mRNA | AK000836.1 | - | 4 | 413 | ||
BC033206.1 | - | 4 | 422 | |||
D26488.1 | - | 5 | 690 | |||
D87716.1 | - | 3 | 196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
