siRNA Details
Gene Symbol: WDR43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015131.2 | NP_055946.1 | 8 | 1122 | ||
GenBank mRNA | AK000836.1 | - | 7 | 818 | ||
BC033206.1 | - | 7 | 827 | |||
D26488.1 | - | 8 | 1095 | |||
D87716.1 | - | 6 | 601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
