siRNA Details
Gene Symbol: LRRC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073385.1 | 3 | 311 | |||
GenBank mRNA | AJ312775.1 | - | 1 | 1969 | ||
BX279889.1 | - | 3 | 305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
