siRNA Details
Gene Symbol: NBEAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015175.2 | NP_055990.1 | 13 | 1748 | ||
XM_006713072.3 | XP_006713135.1 | 12 | 1664 | |||
XM_011533533.2 | XP_011531835.1 | 13 | 1745 | |||
XM_017006010.1 | XP_016861499.1 | 13 | 1745 | |||
XM_017006011.1 | XP_016861500.1 | 13 | 1601 | |||
XM_017006012.1 | XP_016861501.1 | 12 | 1664 | |||
XM_017006013.1 | XP_016861502.1 | 13 | 1745 | |||
XM_017006014.1 | XP_016861503.1 | 12 | 1520 | |||
XM_017006015.1 | XP_016861504.1 | 11 | 1376 | |||
XM_017006016.1 | XP_016861505.1 | 8 | 2322 | |||
GenBank mRNA | AK092871.1 | - | 12 | 1521 | ||
AK131104.1 | - | 13 | 1588 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
