siRNA Details
Gene Symbol: IGF2BP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126326.1 | 1 | 224 | |||
GenBank mRNA | AK057462.1 | - | 1 | 184 | ||
BC041450.1 | - | 1 | 224 | |||
BC137313.1 | - | 1 | 67 | |||
BC137314.1 | - | 1 | 67 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
