siRNA Details
Gene Symbol: UBXN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015562.1 | NP_056377.1 | 7 | 716 | ||
XM_011512670.2 | XP_011510972.1 | 7 | 664 | |||
XM_011512671.2 | XP_011510973.1 | 6 | 521 | |||
GenBank mRNA | AB018337.1 | - | 7 | 644 | ||
AK026451.1 | - | 7 | 669 | |||
AK303630.1 | - | 6 | 521 | |||
AK307899.1 | - | 3 | 277 | |||
BC028986.1 | - | 6 | 548 | |||
BC047420.1 | - | 7 | 666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
