siRNA Details
Gene Symbol: UBXN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015562.1 | NP_056377.1 | 11 | 2114 | ||
XM_011512670.2 | XP_011510972.1 | 11 | 2062 | |||
XM_011512671.2 | XP_011510973.1 | 10 | 1919 | |||
GenBank mRNA | AB018337.1 | - | 11 | 2042 | ||
AK129880.1 | - | 8 | 712 | |||
BC028986.1 | - | 10 | 1947 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
