siRNA Details
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 1017 | |||
NR_136193.1 | 4 | 1714 | ||||
NR_136194.1 | 4 | 1184 | ||||
NR_136197.1 | 4 | 1088 | ||||
NR_136198.1 | 3 | 905 | ||||
NR_136199.1 | 5 | 1227 | ||||
NR_136200.1 | 4 | 1717 | ||||
NR_136201.1 | 4 | 1884 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 1625 | ||
AK021601.1 | - | 1 | 1364 | |||
AK300846.1 | - | 4 | 1095 | |||
EU000853.1 | - | 4 | 930 | |||
EU000854.1 | - | 4 | 930 | |||
EU000855.1 | - | 3 | 2022 | |||
EU000856.1 | - | 3 | 2022 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
