siRNA Details
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 927 | |||
NR_136193.1 | 4 | 1624 | ||||
NR_136194.1 | 4 | 1094 | ||||
NR_136197.1 | 4 | 998 | ||||
NR_136198.1 | 3 | 815 | ||||
NR_136199.1 | 5 | 1137 | ||||
NR_136200.1 | 4 | 1627 | ||||
NR_136201.1 | 4 | 1794 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 1535 | ||
AK021601.1 | - | 1 | 1274 | |||
AK300846.1 | - | 4 | 1005 | |||
EU000853.1 | - | 4 | 840 | |||
EU000854.1 | - | 4 | 840 | |||
EU000855.1 | - | 3 | 1932 | |||
EU000856.1 | - | 3 | 1932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
