siRNA Details
Gene Symbol: ICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015325.2 | NP_056140.1 | 13 | 2441 | ||
XM_011513999.2 | XP_011512301.1 | 13 | 2441 | |||
XM_017009285.1 | XP_016864774.1 | 14 | 2530 | |||
GenBank mRNA | AB023164.3 | - | 13 | 2420 | ||
AK096615.1 | - | 1 | 671 | |||
AL832204.1 | - | 1 | 912 | |||
CR749441.1 | - | 6 | 1712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
