siRNA Details
Gene Symbol: ELOVL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104558.1 | NP_001098028.1 | 8 | 1960 | ||
NM_001297617.1 | NP_001284546.1 | 11 | 2358 | |||
NM_001297618.1 | NP_001284547.1 | 9 | 1965 | |||
NM_024930.2 | NP_079206.2 | 9 | 2011 | |||
XM_005248606.4 | XP_005248663.1 | 8 | 2575 | |||
XM_005248607.3 | XP_005248664.1 | 9 | 2288 | |||
XM_006714695.3 | XP_006714758.1 | 10 | 2067 | |||
XM_011543651.2 | XP_011541953.1 | 10 | 2257 | |||
XM_017009885.1 | XP_016865374.1 | 9 | 1948 | |||
XM_017009886.1 | XP_016865375.1 | 11 | 2187 | |||
XM_017009887.1 | XP_016865376.1 | 11 | 2378 | |||
GenBank mRNA | AB181393.1 | - | 9 | 1952 | ||
AK027216.1 | - | 7 | 1781 | |||
AL137506.1 | - | 4 | 1442 | |||
BC094792.1 | - | 11 | 2299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
