siRNA Details
Gene Symbol: C5orf63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164478.1 | NP_001157950.1 | 4 | 306 | ||
NM_001164479.1 | NP_001157951.1 | 4 | 306 | |||
XM_017009464.1 | XP_016864953.1 | 3 | 290 | |||
XM_017009465.1 | XP_016864954.1 | 3 | 290 | |||
GenBank mRNA | AK126569.1 | - | 5 | 404 | ||
DA507232.1 | - | 4 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
