siRNA Details
Gene Symbol: CFAP97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292033.1 | NP_001278962.1 | 2 | 583 | ||
NM_020827.2 | NP_065878.1 | 2 | 470 | |||
XM_017008483.1 | XP_016863972.1 | 2 | 728 | |||
XM_017008484.1 | XP_016863973.1 | 2 | 362 | |||
XM_017008485.1 | XP_016863974.1 | 4 | 595 | |||
XM_017008486.1 | XP_016863975.1 | 2 | 362 | |||
GenBank mRNA | AB037851.1 | - | 1 | 310 | ||
AK091993.1 | - | 2 | 401 | |||
BC020772.1 | - | 2 | 408 | |||
BC030535.1 | - | 2 | 582 | 5240222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
