siRNA Details
Gene Symbol: CFAP97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020827.2 | NP_065878.1 | 5 | 1651 | ||
XM_017008483.1 | XP_016863972.1 | 5 | 1909 | |||
XM_017008484.1 | XP_016863973.1 | 5 | 1543 | |||
XM_017008485.1 | XP_016863974.1 | 7 | 1776 | |||
XM_017008486.1 | XP_016863975.1 | 5 | 1543 | |||
GenBank mRNA | AB037851.1 | - | 4 | 1491 | ||
AK091993.1 | - | 5 | 1582 | |||
AL133615.1 | - | 4 | 619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
