siRNA Details
Gene Symbol: ARHGEF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080479.2 | NP_001073948.2 | 15 | 2043 | ||
NM_001177693.1 | NP_001171164.1 | 15 | 2043 | |||
NM_001244364.1 | NP_001231293.1 | 7 | 999 | |||
XM_005248568.4 | XP_005248625.1 | 7 | 2480 | |||
XM_011543546.1 | XP_011541848.1 | 15 | 2050 | |||
XM_011543547.2 | XP_011541849.1 | 14 | 2277 | |||
XM_017009702.1 | XP_016865191.1 | 14 | 1728 | |||
XM_017009703.1 | XP_016865192.1 | 6 | 781 | |||
GenBank mRNA | AK094713.1 | - | 15 | 2043 | ||
AK304761.1 | - | 7 | 999 | |||
BC157846.1 | - | 14 | 1889 | 9057065 | ||
BC171850.1 | - | 14 | 1889 | 9054504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
