siRNA Details
Gene Symbol: TXLNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153235.3 | NP_694967.3 | 10 | 3580 | ||
XM_005266836.1 | XP_005266893.1 | 11 | 3947 | |||
XM_011535505.2 | XP_011533807.1 | 10 | 3684 | |||
XM_011535506.2 | XP_011533808.1 | 9 | 3130 | |||
XM_017010322.1 | XP_016865811.1 | 11 | 3810 | |||
GenBank mRNA | AF111704.1 | - | 1 | 1526 | ||
AK056970.1 | - | 1 | 721 | |||
AL831988.1 | - | 10 | 3518 | |||
AL832322.1 | - | 7 | 2878 | |||
AL834248.2 | - | 10 | 3509 | |||
BX647477.1 | - | 10 | 3528 | |||
BX647633.1 | - | 10 | 3503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
