siRNA Details
Gene Symbol: TXLNB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153235.3 | NP_694967.3 | 10 | 3648 | ||
XM_005266836.1 | XP_005266893.1 | 11 | 4015 | |||
XM_011535505.2 | XP_011533807.1 | 10 | 3752 | |||
XM_011535506.2 | XP_011533808.1 | 9 | 3198 | |||
XM_017010322.1 | XP_016865811.1 | 11 | 3878 | |||
GenBank mRNA | AF111704.1 | - | 1 | 1594 | ||
AK056970.1 | - | 1 | 789 | |||
AL831988.1 | - | 10 | 3586 | |||
AL832322.1 | - | 7 | 2946 | |||
AL834248.2 | - | 10 | 3577 | |||
BX647477.1 | - | 10 | 3596 | |||
BX647633.1 | - | 10 | 3571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
