siRNA Details
Gene Symbol: C7orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161834.2 | NP_001155306.2 | 9 | 1219 | ||
XM_011515052.1 | XP_011513354.1 | 8 | 1402 | |||
GenBank mRNA | DY654485.1 | - | 2 | 158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
