siRNA Details
Gene Symbol: HOXA-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122069.1 | 1 | 52 | |||
GenBank mRNA | AK311383.1 | - | 2 | 309 | ||
HY010098.1 | - | 4 | 403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
