siRNA Details
Gene Symbol: GRID2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145118.1 | NP_001138590.1 | 18 | 3124 | ||
XM_011515404.2 | XP_011513706.1 | 17 | 2599 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
